accessibility menu, dialog, popup

UserName

CAS RN 52109-66-7

IUPAC Name: 5-phenylpyrazine-2,3-dicarbonitrile
Synonyms: 5-phenylpyrazine-2,3-dicarbonitrile
Molecular Weight (g/mol): 206.21
Molecular Formula: C12H6N4
InChi Key: KSVMJUBGOLBDJX-UHFFFAOYSA-N
SMILES: N#CC1=NC=C(N=C1C#N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results