Learn More
2,3-Dicyano-5-phenylpyrazine, 97%, Thermo Scientific™
Product Code. 15494618
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15494618 2 g 2g
15404628 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15494618 Supplier Thermo Scientific Alfa Aesar Supplier No. H58294.04
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 52109-66-7
Molecular Formula C12H6N4
Molecular Weight (g/mol) 206.208
MDL Number MFCD03081218
InChI Key KSVMJUBGOLBDJX-UHFFFAOYSA-N
Synonym 2,3-dicyano-5-phenylpyrazine, 5-phenyl-2,3-pyrazinedicarbonitrile, 2,3-pyrazinedicarbonitrile,5-phenyl, 5-phenyl-pyrazine-2,3-dicarbonitrile, 5-phenyl-2,3-pyrazinedicarbonitrile #
PubChem CID 142907
IUPAC Name 5-phenylpyrazine-2,3-dicarbonitrile
SMILES C1=CC=C(C=C1)C2=CN=C(C(=N2)C#N)C#N
Specifications
Quantity 2 g
UN Number UN3439
Formula Weight 206.21
Percent Purity 97%
Chemical Name or Material 2,3-Dicyano-5-phenylpyrazine