accessibility menu, dialog, popup

UserName

N(S)OHNOHOOCH3CH3CH3O

CAS RN 47355-10-2

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-(1-formyl-1H-indol-3-yl)propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-(1-formylindol-3-yl)propanoic acid
Molecular Weight (g/mol): 332.36
Molecular Formula: C17H20N2O5
InChi Key: IHXHBYFWSOYYTR-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CN(C=O)C2=CC=CC=C12)C(O)=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results