missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1-formyl-L-tryptophan, 98%, Thermo Scientific™

Product Code. 15430229
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430229 100 g 100g
15410229 5 g 5g
15420229 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15430229 Supplier Thermo Scientific Alfa Aesar Supplier No. H63499.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 47355-10-2
Molecular Formula C17H20N2O5
Molecular Weight (g/mol) 332.36
MDL Number MFCD00065992
InChI Key IHXHBYFWSOYYTR-ZDUSSCGKSA-N
Synonym boc-trp for-oh, n-tert-butoxycarbonyl-n'-formyl-l-tryptophan, chembl63415, nalpha-tert-butoxycarbonyl-n1-formyl-l-tryptophan, s-2-tert-butoxycarbonyl amino-3-1-formyl-1h-indol-3-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-formylindol-3-yl propanoic acid, boc-trp cho-oh, pubchem12191, t-boc-trp cho-oh, nalpha-boc-n1-formyl-l-tryptophan
PubChem CID 7017963
IUPAC Name (2S)-3-(1-formylindol-3-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CN(C=O)C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 100°C (decomposition)
Beilstein 4299614
Quantity 100 g
Formula Weight 332.35
Sensitivity Light sensitive
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-1-formyl-L-tryptophan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.