accessibility menu, dialog, popup

UserName

CAS RN 464-48-2

H3CH3C(S)CH3O

CAS RN 464-48-2

IUPAC Name: (1S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
Synonyms: (1S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: DSSYKIVIOFKYAU-OMNKOJBGSA-N
SMILES: CC1(C)C2CC[C@]1(C)C(=O)C2
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)