Learn More
L-Camphor, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
L-Camphor
Specifications
Specifications
| Chemical Name or Material | (1S)-(-)-Camphor |
| CAS | 464-48-2 |
| Analyte or Component Names | (1S)-(-)-Camphor |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H16 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | (1S,4S)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, Bicyclo[2.2.1]heptan-2-one, 1,7,7-trimethyl-, (1S)-, Camphor, (1S,4S)-(-)- (8CI), (-)-Alcanfor, (-)-Camphor, (1S)-(-)-Camphor, (1S)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (1S)-Camphor, (1S,4S)-(-)-Camphor, (S)-(-)-Camphor, NSC 26351, l-Camphor, Bicyclo[2.2.1]heptan-2-one, 1,7,7-trimethyl-, (1S,4S)- |
| InChI Formula | InChI=1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m0/s1 |
| SMILES | CC1(C)[C@H]2CC[C@]1(C)C(=O)C2 |
| IUPAC Name | (1S,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one |
| Show More |