accessibility menu, dialog, popup

UserName

H3CSiH3CCH3NNN

CAS RN 43183-36-4

IUPAC Name: 1-(trimethylsilyl)-1H-1,2,3-benzotriazole
Synonyms: 1-(trimethylsilyl)-1,2,3-benzotriazole
Molecular Weight (g/mol): 191.31
Molecular Formula: C9H13N3Si
InChi Key: VHOLOZYRMQVHBK-UHFFFAOYSA-N
SMILES: C[Si](C)(C)N1N=NC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)