Learn More
1-(Trimethylsilyl)-1H-benzotriazole, TRC
Product Code. 30452651
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452651 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30452651 Supplier Toronto Research Chemicals Supplier No. TRCT798323500MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(Trimethylsilyl)-1H-benzotriazole
CAS 43183-36-4
Molecular Formula C9H13N3Si
Quantity 500 mg
InChI Formula InChI=1S/C9H13N3Si/c1-13(2,3)12-9-7-5-4-6-8(9)10-11-12/h4-7H,1-3H3
SMILES C[Si](C)(C)n1nnc2ccccc12
IUPAC Name benzotriazol-1-yl(trimethyl)silane
Molecular Weight (g/mol) 191.3
Formula Weight 191.0879