Learn More
1-(Trimethylsilyl)-1H-benzotriazole, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(Trimethylsilyl)-1H-benzotriazole |
| CAS | 43183-36-4 |
| Molecular Formula | C9H13N3Si |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C9H13N3Si/c1-13(2,3)12-9-7-5-4-6-8(9)10-11-12/h4-7H,1-3H3 |
| SMILES | C[Si](C)(C)n1nnc2ccccc12 |
| IUPAC Name | benzotriazol-1-yl(trimethyl)silane |
| Molecular Weight (g/mol) | 191.3 |
| Formula Weight | 191.0879 |