accessibility menu, dialog, popup

UserName

NHOCl

CAS RN 40673-25-4

IUPAC Name: 4-chloro-1,2-dihydropyridin-2-one
Synonyms: 4-chloro-1H-pyridin-2-one
Molecular Weight (g/mol): 129.54
Molecular Formula: C5H4ClNO
InChi Key: OZUXGFRLSKQVMI-UHFFFAOYSA-N
SMILES: ClC1=CC(=O)NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results