missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-hydroxypyridine, 98%

Product Code. 15473847
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15473847 250 mg 250mg
11482280 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473847 Supplier Thermo Scientific Alfa Aesar Supplier No. H29250.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 40673-25-4 | C5H4ClNO | 129.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40673-25-4
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.54
MDL Number MFCD03093022
InChI Key OZUXGFRLSKQVMI-UHFFFAOYSA-N
Synonym 4-chloro-2-hydroxypyridine, 4-chloropyridin-2-ol, 4-chloro-2-pyridinol, 2 1h-pyridinone, 4-chloro, 4-chloro-2 1h-pyridinone, 4-chloro-2 1h-pyridone, 4-chloropyridone, 4-chloro pyridone, 4-chloro-2-pyridone, pubchem9621
PubChem CID 2757611
IUPAC Name 4-chloro-1H-pyridin-2-one
SMILES ClC1=CC(=O)NC=C1

Specifications

Melting Point 177°C to 179°C
Quantity 250 mg
Formula Weight 129.55
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-hydroxypyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.