accessibility menu, dialog, popup

UserName

H3CON

CAS RN 36263-51-1

IUPAC Name: 1-(4-methoxyphenyl)cyclohexane-1-carbonitrile
Synonyms: 1-(4-methoxyphenyl)cyclohexane-1-carbonitrile
Molecular Weight (g/mol): 215.3
Molecular Formula: C14H17NO
InChi Key: YQXUSXSQFVWPRQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1(CCCCC1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results