missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methoxyphenyl)-1-cyclohexanecarbonitrile, 94%, Thermo Scientific™

Product Code. 11382376
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382376 25 g 25g
11372376 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382376 Supplier Thermo Scientific Alfa Aesar Supplier No. L06107.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36263-51-1
Molecular Formula C14H17NO
Molecular Weight (g/mol) 215.296
MDL Number MFCD00021282
InChI Key YQXUSXSQFVWPRQ-UHFFFAOYSA-N
PubChem CID 98629
IUPAC Name 1-(4-methoxyphenyl)cyclohexane-1-carbonitrile
SMILES COC1=CC=C(C=C1)C2(CCCCC2)C#N

Specifications

Melting Point 43°C to 60°C
Quantity 25 g
UN Number UN3439
Beilstein 2941304
Synonym 1-4-methoxyphenyl cyclohexanecarbonitrile, 1-4-methoxyphenyl-1-cyclohexanecarbonitrile, 1-4-methoxyphenyl cyclohexane-1-carbonitrile, cyclohexanecarbonitrile, 1-4-methoxyphenyl, acmc-20apah, 4-1-cyanocyclohexyl anisole, 1-4-methoxyphenyl cyclohexanecarbonitrile #, cyclohexanecarbonitrile,1-4-methoxyphenyl
Formula Weight 215.3
Percent Purity 94%
Chemical Name or Material 1-(4-Methoxyphenyl)-1-cyclohexanecarbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.