accessibility menu, dialog, popup

UserName

CAS RN 3301-94-8

IUPAC Name: 6-butyloxan-2-one
Synonyms: delta-nonalactone
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: PXRBWNLUQYZAAX-UHFFFAOYNA-N
SMILES: CCCCC1CCCC(=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results