Learn More
delta-Nonanolactone, 98%, Thermo Scientific Chemicals
Product Code. 11358985
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358985 100 g 100g
11348985 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11358985 Supplier Thermo Scientific Alfa Aesar Supplier No. L14122.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identificadores de productos químicos
CAS 3301-94-8
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00036500
InChI Key PXRBWNLUQYZAAX-UHFFFAOYSA-N
Synonym delta-nonalactone, 5-nonanolide, 6-butyltetrahydro-2h-pyran-2-one, 2h-pyran-2-one, 6-butyltetrahydro, 5-nonalactone, delta-nonanolactone, hydroxynonanoic acid delta-lactone, 5-butyl-delta-valerolactone, delta-butyl-delta-valerolactone, delta-hydroxypelargonic acid lactone
PubChem CID 18698
IUPAC Name 6-butyloxan-2-one
SMILES CCCCC1CCCC(=O)O1
Especificaciones
Melting Point -26°C
Density 0.99
Boiling Point 250°C
Flash Point 135°C (275°F)
Refractive Index 1.457
Quantity 100 g
Beilstein 114460
Formula Weight 156.23
Percent Purity 98%
Chemical Name or Material delta-Nonanolactone
RUO – Research Use Only
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.