accessibility menu, dialog, popup

UserName

FFFBrCl

CAS RN 261763-23-9

IUPAC Name: 4-(bromomethyl)-1-chloro-2-(trifluoromethyl)benzene
Synonyms: 4-(bromomethyl)-1-chloro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 273.48
Molecular Formula: C8H5BrClF3
InChi Key: LZLIPLUATRVXSB-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(CBr)=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results