missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™

Product Code. 11386979
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11386979 5 g 5g
11376979 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386979 Supplier Thermo Scientific Alfa Aesar Supplier No. H26834.06
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261763-23-9
Molecular Formula C8H5BrClF3
Molecular Weight (g/mol) 273.477
MDL Number MFCD01631540
InChI Key LZLIPLUATRVXSB-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl benzyl bromide, 4-bromomethyl-1-chloro-2-trifluoromethyl benzene, 3-trifluoromethyl-4-chlorobenzyl bromide, 4-bromomethyl-1-chloro-2-trifluoromethyl-benzene, benzene, 4-bromomethyl-1-chloro-2-trifluoromethyl, acmc-20amlg, pubchem3030, 3-trifluoromethyl-4-chlorobenzylbromide, 3-trifluromethyl-4-chlorobenzyl bromide, 4-chloro-3-trifluoromethylbenzyl bromide
PubChem CID 2778114
IUPAC Name 4-(bromomethyl)-1-chloro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1CBr)C(F)(F)F)Cl

Specifications

Melting Point 36°C to 40°C
Quantity 5 g
UN Number UN3261
Formula Weight 273.48
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.