accessibility menu, dialog, popup

UserName

ClSSCl

CAS RN 2393-97-7

IUPAC Name: 1-chloro-4-({[(4-chlorophenyl)sulfanyl]methyl}sulfanyl)benzene
Synonyms: 1-chloro-4-({[(4-chlorophenyl)sulfanyl]methyl}sulfanyl)benzene
Molecular Weight (g/mol): 301.24
Molecular Formula: C13H10Cl2S2
InChi Key: NTZBITFHYUVXFR-UHFFFAOYSA-N
SMILES: ClC1=CC=C(SCSC2=CC=C(Cl)C=C2)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results