accessibility menu, dialog, popup

UserName

OHNSBr(E)

CAS RN 2160-63-6

IUPAC Name: (E)-N-[(5-bromothiophen-2-yl)methylidene]hydroxylamine
Synonyms: (E)-N-[(5-bromothiophen-2-yl)methylidene]hydroxylamine
Molecular Weight (g/mol): 206.06
Molecular Formula: C5H4BrNOS
InChi Key: URLSTLFAZRSULI-XVNBXDOJSA-N
SMILES: O\N=C\C1=CC=C(Br)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results