missing translation for 'onlineSavingsMsg'
Learn More

5-Bromothiophene-2-carboxaldoxime, 97%, Thermo Scientific™

Product Code. 11451527
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11451527 5 g 5g
11441527 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11451527 Supplier Thermo Scientific Alfa Aesar Supplier No. B24968.06
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2160-63-6
Molecular Formula C5H4BrNOS
Molecular Weight (g/mol) 206.06
MDL Number MFCD00126375
InChI Key URLSTLFAZRSULI-XVNBXDOJSA-N
Synonym 5-bromothiophene-2-carboxaldehyde oxime, 5-bromothiophene-2-carbaldehyde oxime, 5-bromo-2-thiophenaldoxime, 5-bromothiophene-2-carboxaldoxime, 5-bromo-2-thiophenecarboxaldehyde oxime, 2-thiophenecarboxaldehyde, 5-bromo-, oxime, 5-bromo-thiophene-2-carbaldehyde oxime, 5-bromo 2-thienyl hydroxyimino methane, 2e-5-bromo-2-thiophenecarboxaldehyde oxime, e-1-5-bromothiophen-2-yl-n-hydroxymethanimine
PubChem CID 9578377
IUPAC Name (NE)-N-[(5-bromothiophen-2-yl)methylidene]hydroxylamine
SMILES O\N=C\C1=CC=C(Br)S1

Specifications

Melting Point 132°C to 134°C
Quantity 5 g
Formula Weight 206.06
Percent Purity 97%
Chemical Name or Material 5-Bromothiophene-2-carboxaldoxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.