accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3COSiCH3H3CCH3

CAS RN 2096331-55-2

IUPAC Name: trimethyl({2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)propyl]phenoxy})silane
Synonyms: trimethyl(2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)propyl]phenoxy)silane
Molecular Weight (g/mol): 334.34
Molecular Formula: C18H31BO3Si
InChi Key: WVIXWRBQCZVWHF-UHFFFAOYSA-N
SMILES: CC1(C)OB(CCCC2=CC=CC=C2O[Si](C)(C)C)OC1(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results