missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Trimethylsiloxyphenyl)-1-propylboronic acid pinacol ester, 95%, Thermo Scientific™

Product Code. 15497755
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497755 1 g 1g
15407765 5 g 5g
15417765 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15497755 Supplier Thermo Scientific Alfa Aesar Supplier No. H30323.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2096331-55-2
Molecular Formula C18H31BO3Si
Molecular Weight (g/mol) 334.34
MDL Number MFCD12546185
InChI Key WVIXWRBQCZVWHF-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-3-2-trimethylsiloxyphenyl propyl-1,3,2-dioxaborolane, 3-2-trimethylsiloxyphenyl-1-propylboronic acid pinacol ester
PubChem CID 73995112

Specifications

Refractive Index 1.4799
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 334.34
Percent Purity 95%
Chemical Name or Material 3-(2-Trimethylsiloxyphenyl)-1-propylboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.