accessibility menu, dialog, popup

UserName

FNNOHNF

CAS RN 2062-78-4

IUPAC Name: 1-{1-[4,4-bis(4-fluorophenyl)butyl]piperidin-4-yl}-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: pimozide orap asta medica brand of pimozide pimozide (substance) janssen brand of pimozide antalon pimozidum opiran pharmascience brand of pimozide Orap
Molecular Weight (g/mol): 461.56
Molecular Formula: C28H29F2N3O
InChi Key: YVUQSNJEYSNKRX-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(CCCN1CCC(CC1)N1C(=O)NC2=CC=CC=C12)C1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (7)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (7)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)