CAS RN 192-97-2
CAS RN 192-97-2
IUPAC Name:
benzo[e]pyrene
Synonyms:
benzo(E)pyrene benz(e)pyrene 4,5-benzopyrene 4,5-benzpyrene benzo(l)pyrene 1,2-benzopyrene 1,2-benzo(e)pyrene B(e)P ccris 786 1,2-benzpyrene
Molecular Weight (g/mol):
252.32
Molecular Formula:
C20H12
InChi Key:
TXVHTIQJNYSSKO-UHFFFAOYSA-N
SMILES:
C1=CC=C2C(=C1)C1=CC=CC3=CC=C4C=CC=C2C4=C13