Learn More
Benzo(e)pyrene in Acetonitrile 10μg/mL, Fisher Chemical™
Product Code. 12929633 Shop All Fisher Chemical Products
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Product Code. 12929633
Brand: Fisher Chemical P806890
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Ampoule(s)
CAS 192-97-2
Molecular Formula C20H12
Concentration 10 μg/mL
UN Number 1648
InChI Key TXVHTIQJNYSSKO-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C3=CC=CC4=C3C5=C(C=CC=C25)C=C4
IUPAC Name benzo[e]pyrene
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 252.316
PubChem CID 9128
Quantity 10 mL
ChEBI CHEBI:34567
Chemical Name or Material Benzo(e)pyrene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Benzo(e)pyrene 10ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .