Learn More
Benzo(e)pyrene, SPEX CertiPrep™
Product Code. 12929434 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 12929434
Brand: SPEX Certiprep S440
Specifications
Packaging Ampule
CAS 192-97-2, 75-09-2
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
Molecular Formula C20H12
InChI Key TXVHTIQJNYSSKO-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C3=CC=CC4=C3C5=C(C=CC=C25)C=C4
IUPAC Name benzo[e]pyrene
Molecular Weight (g/mol) 252.316
PubChem CID 9128
Quantity 1 mL
ChEBI CHEBI:34567
Chemical Name or Material Benzo(e)pyrene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzo(e)pyrene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H350-May cause cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : dichloromethane, Benzo[e]pyrene