accessibility menu, dialog, popup

UserName

CAS RN 1738-78-9

IUPAC Name: 4-methylbenzene-1-sulfonic acid; benzyl 2-amino-3-phenylpropanoate
Synonyms: benzyl 2-amino-3-phenylpropanoate; para-toluene sulfonate
Molecular Weight (g/mol): 427.52
Molecular Formula: C23H25NO5S
InChi Key: ZLZGBBIPWXUQST-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.NC(CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results