missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine benzyl ester p-toluenesulfonate, 95%, Thermo Scientific™

Product Code. 15427756
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15427756 25 g 25g
15417756 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15427756 Supplier Thermo Scientific Alfa Aesar Supplier No. H62700.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1738-78-9
Molecular Formula C23H25NO5S
Molecular Weight (g/mol) 427.515
MDL Number MFCD00035105
InChI Key ZLZGBBIPWXUQST-RSAXXLAASA-N
Synonym h-phe-obzl p-tosylate, l-phenylalanine benzyl ester p-toluenesulfonate, 3-phenyl-l-alanine benzyl ester 4-toluenesulphonate, h-phe-obzl.tosoh, l-phenylalanine benzyl ester tosylate, s-benzyl 2-amino-3-phenylpropanoate 4-methylbenzenesulfonate, o-benzyl-3-phenyl-l-alanine toluene-4-sulphonate, benzyl 2s-2-amino-3-phenylpropanoate; para-toluene sulfonate, h-phe-obzl tos, h-phe-obzl. tos
PubChem CID 12636207
IUPAC Name benzyl (2S)-2-amino-3-phenylpropanoate;4-methylbenzenesulfonic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.C1=CC=C(C=C1)CC(C(=O)OCC2=CC=CC=C2)N

Specifications

Quantity 25 g
Formula Weight 427.51
Percent Purity 95%
Chemical Name or Material L-Phenylalanine benzyl ester p-toluenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.