accessibility menu, dialog, popup

UserName

CAS RN 14906-64-0

ONHCHN

CAS RN 14906-64-0

IUPAC Name: 3-cyano-1,4-dihydro-1λ²-pyridine-1,1-diium-4-id-1-olate
Synonyms: 3-cyano-1,4-dihydro-1λ²-pyridine-1,1-diium-4-id-1-olate
Molecular Weight (g/mol): 121.12
Molecular Formula: C6H5N2O
InChi Key: AABSWFBNBQBPEQ-UHFFFAOYNA-N
SMILES: [O-][NH++]1C=C[CH-]C(=C1)C#N
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)