Learn More
3-Cyanopyridine 1-oxide, 97%
Product Code. 11428837
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428837 10 g 10g
11418837 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11428837 Supplier Thermo Scientific Alfa Aesar Supplier No. A19961.09
Request Bulk or Custom Format
CAS: 14906-64-0 | C6H5N2O | 121.12 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14906-64-0
Molecular Formula C6H5N2O
Molecular Weight (g/mol) 121.12
MDL Number MFCD00129037
InChI Key AABSWFBNBQBPEQ-UHFFFAOYNA-N
Synonym nicotinonitrile-1-oxide, 3-cyanopyridine n-oxide, 3-cyanopyridine-n-oxide, nicotinonitrile 1-oxide, 3-cyanopyridine 1-oxide, 3-cyanopyridin-1-ium-1-olate, 3-pyridinecarbonitrile, 1-oxide, acmc-1c6hr, 3-pyridinecarbonitrile,1-oxide, woovsqcalyyudo-uhfffaoysa
PubChem CID 84686
SMILES [O-][NH++]1C=C[CH-]C(=C1)C#N
Specifications
Melting Point 175°C to 178°C
Quantity 10 g
Sensitivity Hygroscopic
IUPAC Name 3-cyano-1,4-dihydro-1λ²-pyridine-1,1-diium-4-id-1-olate
Formula Weight 120.11
Percent Purity 97%
Chemical Name or Material 3-Cyanopyridine 1-oxide