Learn More
3-Cyanopyridine N-Oxide, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Cyanopyridine N-Oxide |
| CAS | 14906-64-0 |
| Molecular Formula | C6H4N2O |
| Quantity | 2.5 g |
| Synonym | 3-Pyridinecarbonitrile 1-Oxide, Nicotinonitrile 1-Oxide, 3-Cyanopyridine 1-Oxide, NSC 170850, Nicotinonitrile N-Oxide |
| InChI Formula | InChI=1S/C6H4N2O/c7-4-6-2-1-3-8(9)5-6/h1-3,5H |
| SMILES | O=N1=CC(C#N)=CC=C1 |
| IUPAC Name | 3-cyanopyridine 1-oxide |
| Molecular Weight (g/mol) | 120.11 |
| Formula Weight | 120.03 |