accessibility menu, dialog, popup

UserName

H3COOOOOH3COOCH3OOH3C

CAS RN 144490-03-9

IUPAC Name: [3,4,5-tris(acetyloxy)oxolan-2-yl]methyl acetate
Synonyms: [3,4,5-tris(acetyloxy)oxolan-2-yl]methyl acetate
Molecular Weight (g/mol): 318.28
Molecular Formula: C13H18O9
InChi Key: IHNHAHWGVLXCCI-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results