Learn More
1,2,3,5-Tetra-O-acetyl beta-L-Ribofuranose, TRC
Product Code. 30418344
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418344 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30418344 Supplier Toronto Research Chemicals Supplier No. TRCT283105500MG
 Restricted Item
Estimated Shipment: 24-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,2,3,5-Tetra-O-acetyl beta-L-Ribofuranose
CAS 144490-03-9
Molecular Formula C13 H18 O13
Quantity 500 mg
Synonym 1,2,3,5-Tetra-O-acetyl β-L-Ribofuranose, β-L-Ribofuranose Tetraacetate
InChI Formula InChI=1S/C13H18O9/c1-6(14)18-5-10-11(19-7(2)15)12(20-8(3)16)13(22-10)21-9(4)17/h10-13H,5H2,1-4H3/t10-,11-,12-,13-/m0/s1
SMILES CC(=O)OC[C@@H]1O[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@H]1OC(=O)C
IUPAC Name [(2S,3S,4S,5R)-3,4,5-triacetoxytetrahydrofuran-2-yl]methyl acetate
Molecular Weight (g/mol) 382.27
Formula Weight 382.0747