Learn More
1,2,3,5-Tetra-O-acetyl beta-L-Ribofuranose, TRC
Product Code. 30424117
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424117 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30424117 Supplier Toronto Research Chemicals Supplier No. TRCT2831051G
 Restricted Item
Estimated Shipment: 24-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,2,3,5-Tetra-O-acetyl beta-L-Ribofuranose
CAS 144490-03-9
Molecular Formula C13 H18 O13
Quantity 1 g
Synonym 1,2,3,5-Tetra-O-acetyl β-L-Ribofuranose, β-L-Ribofuranose Tetraacetate
InChI Formula InChI=1S/C13H18O9/c1-6(14)18-5-10-11(19-7(2)15)12(20-8(3)16)13(22-10)21-9(4)17/h10-13H,5H2,1-4H3/t10-,11-,12-,13-/m0/s1
SMILES CC(=O)OC[C@@H]1O[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@H]1OC(=O)C
IUPAC Name [(2S,3S,4S,5R)-3,4,5-triacetoxytetrahydrofuran-2-yl]methyl acetate
Molecular Weight (g/mol) 382.27
Formula Weight 382.0747