CAS RN 1373431-65-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Talazoparib Tosylate, TRC
CAS: 1373431-65-2 Molecular Formula: C19H14F2N6O . C7H8O3S Molecular Weight (g/mol): 552.6 Synonym: BMN 673ts; SMILES: Cc1ccc(cc1)S(=O)(=O)O.Cn2ncnc2[C@@H]3[C@H](Nc4cc(F)cc5C(=O)NN=C3c45)c6ccc(F)cc6
2
Talazoparib tosylate, MedChemExpress
MedChemExpress Talazoparib tosylate (BMN 673ts) is a novel, potent and orally available PARP1/2 inhibitor with an IC50 of 0.57 nM for PARP1.