Learn More
Talazoparib tosylate, MedChemExpress
Product Code. 16490537
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490537 50 mg 50mg
16486907 5 mg 5mg
16404198 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16490537 Supplier Medchem Express Supplier No. HY108413
MedChemExpress Talazoparib tosylate (BMN 673ts) is a novel, potent and orally available PARP1/2 inhibitor with an IC50 of 0.57 nM for PARP1.
Talazoparib tosylate (BMN 673ts) is a novel, potent and orally available PARP1/2 inhibitor with an IC50 of 0.57 nM for PARP1.
Specifications
Chemical Name or Material Talazoparib tosylate
CAS 1373431-65-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H22F2N6O4S
Quantity 50 mg
Synonym BMN 673ts
Solubility Information DMSO : ≥ 108 mg/mL (195.46 mM)
SMILES O=C1NN=C2C3=C1C=C(F)C=C3N[C@H](C4=CC=C(F)C=C4)[C@H]2C5=NC=NN5C.O=S(C6=CC=C(C)C=C6)(O)=O
Molecular Weight (g/mol) 552.55
Formula Weight 552.55
Percent Purity 99.72%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death