Learn More
Talazoparib Tosylate, TRC
Product Code. 30009093
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009093 50 mg 50mg
30006495 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30009093 Supplier Toronto Research Chemicals Supplier No. TRCT13734050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1373431-65-2
Molecular Formula C19H14F2N6O . C7H8O3S
Molecular Weight (g/mol) 552.6
Synonym BMN 673ts;
SMILES Cc1ccc(cc1)S(=O)(=O)O.Cn2ncnc2[C@@H]3[C@H](Nc4cc(F)cc5C(=O)NN=C3c45)c6ccc(F)cc6
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C19H14F2N6O.C7H8O3S/c1-27-18(22-8-23-27)15-16(9-2-4-10(20)5-3-9)24-13-7-11(21)6-12-14(13)17(15)25-26-19(12)28;1-6-2-4-7(5-3-6)11(8,9)10/h2-8,15-16,24H,1H3,(H,26,28);2-5H,1H3,(H,8,9,10)/t15-,16-;/m1./s1
Formula Weight 552.1391
Chemical Name or Material Talazoparib Tosylate