accessibility menu, dialog, popup

UserName

ClH3CNH(S)OSH

CAS RN 136434-34-9

IUPAC Name: hydrogen methyl[(3S)-3-(naphthalen-1-yloxy)-3-(thiophen-2-yl)propyl]amine chloride
Synonyms: hydrogen duloxetine chloride
Molecular Weight (g/mol): 333.87
Molecular Formula: C18H20ClNOS
InChi Key: BFFSMCNJSOPUAY-LMOVPXPDSA-N
SMILES: [H+].[Cl-].CNCC[C@H](OC1=C2C=CC=CC2=CC=C1)C1=CC=CS1
Industry Type 
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (4)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)