accessibility menu, dialog, popup

UserName

ClOOClClCl

CAS RN 133406-29-8

IUPAC Name: 6-chloro-2-(trichloromethyl)-4H-chromen-4-one
Synonyms: 6-chloro-2-(trichloromethyl)chromen-4-one
Molecular Weight (g/mol): 297.94
Molecular Formula: C10H4Cl4O2
InChi Key: NTDHYMSVCBGQJF-UHFFFAOYSA-N
SMILES: ClC1=CC=C2OC(=CC(=O)C2=C1)C(Cl)(Cl)Cl
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results