CAS RN 125572-93-2
CAS RN 125572-93-2
IUPAC Name:
hydrogen (6S)-6-{propyl[2-(thiophen-2-yl)ethyl]amino}-5,6,7,8-tetrahydronaphthalen-1-ol chloride
Synonyms:
hydrogen neupro chloride
Molecular Weight (g/mol):
351.93
Molecular Formula:
C19H26ClNOS
InChi Key:
CEXBONHIOKGWNU-NTISSMGPSA-N
SMILES:
[H+].[Cl-].CCCN(CCC1=CC=CS1)[C@H]1CCC2=C(O)C=CC=C2C1
1
–
2
of
2
results
1
Tocris Bioscience™ Rotigotine hydrochloride
CAS: 125572-93-2 Molecular Formula: C19H26ClNOS
2
Rotigotine Hydrochloride, MedChemExpress
MedChemExpress Rotigotine Hydrochloride (N-0923 Hydrochloride) is a full agonist of dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor, with Ki of 0.71 nM, 4-15 nM, and 83 nM for the dopamine D3 receptor and D2, D5, D4 receptors, and dopamine D1 receptor.