accessibility menu, dialog, popup

UserName

CAS RN 1234480-84-2

CH3NNNONHNNNONCH3CH3OH3C

CAS RN 1234480-84-2

IUPAC Name: 5-({2-methoxy-4-[4-(4-methylpiperazin-1-yl)piperidine-1-carbonyl]phenyl}amino)-2,9-dimethyl-2,4,6,9-tetraazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Synonyms: 5-({2-methoxy-4-[4-(4-methylpiperazin-1-yl)piperidine-1-carbonyl]phenyl}amino)-2,9-dimethyl-2,4,6,9-tetraazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Molecular Weight (g/mol): 570.7
Molecular Formula: C31H38N8O3
InChi Key: IWMCPJZTADUIFX-UHFFFAOYSA-N
SMILES: COC1=C(NC2=NC=C3N(C)C(=O)C4=CC=CC=C4N(C)C3=N2)C=CC(=C1)C(=O)N1CCC(CC1)N1CCN(C)CC1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results