accessibility menu, dialog, popup

UserName

H3CNH3CONCl

CAS RN 112006-59-4

IUPAC Name: 2-chloro-N-ethyl-N-methylpyridine-3-carboxamide
Synonyms: 2-chloro-N-ethyl-N-methylpyridine-3-carboxamide
Molecular Weight (g/mol): 198.65
Molecular Formula: C9H11ClN2O
InChi Key: OKOCIHPHSMRJDK-UHFFFAOYSA-N
SMILES: CCN(C)C(=O)C1=C(Cl)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results