CAS RN 1092461-33-0
CAS RN 1092461-33-0
IUPAC Name:
2-(3-ethoxy-2,6-difluorophenyl)acetonitrile
Synonyms:
2-(3-ethoxy-2,6-difluorophenyl)acetonitrile
Molecular Weight (g/mol):
197.19
Molecular Formula:
C10H9F2NO
InChi Key:
SNFOWHHWCGVXTQ-UHFFFAOYSA-N
SMILES:
CCOC1=C(F)C(CC#N)=C(F)C=C1
1
–
1
of
1
results
1
3-Ethoxy-2,6-difluorophenylacetonitrile, 97%, Thermo Scientific™
CAS: 1092461-33-0 Molecular Formula: C10H9F2NO Molecular Weight (g/mol): 197.185 MDL Number: MFCD11519375 InChI Key: SNFOWHHWCGVXTQ-UHFFFAOYSA-N Synonym: 3-ethoxy-2,6-difluorophenylacetonitrile,2-3-ethoxy-2,6-difluorophenyl acetonitrile,2-3-ethoxy-2,6-difluorophenyl ethanenitrile,3-ethoxy-2,6-difluorophenylacetonitrile, jrd PubChem CID: 55252374 IUPAC Name: 2-(3-ethoxy-2,6-difluorophenyl)acetonitrile SMILES: CCOC1=C(C(=C(C=C1)F)CC#N)F