Learn More
3-Ethoxy-2,6-difluorophenylacetonitrile, 97%, Thermo Scientific™
Product Code. 15454587
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15454587 250 mg 250mg
15444587 1 g 1g
2 options
Product Code. 15454587
Brand: Thermo Scientific Alfa Aesar H31931.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1092461-33-0
Molecular Formula C10H9F2NO
Molecular Weight (g/mol) 197.185
MDL Number MFCD11519375
InChI Key SNFOWHHWCGVXTQ-UHFFFAOYSA-N
Synonym 3-ethoxy-2,6-difluorophenylacetonitrile, 2-3-ethoxy-2,6-difluorophenyl acetonitrile, 2-3-ethoxy-2,6-difluorophenyl ethanenitrile, 3-ethoxy-2,6-difluorophenylacetonitrile, jrd
PubChem CID 55252374
IUPAC Name 2-(3-ethoxy-2,6-difluorophenyl)acetonitrile
SMILES CCOC1=C(C(=C(C=C1)F)CC#N)F
Specifications
Melting Point 50°C to 53°C
Quantity 250 mg
UN Number UN3439
Formula Weight 197.18
Percent Purity 97%
Chemical Name or Material 3-Ethoxy-2,6-difluorophenylacetonitrile