missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Znt5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25604-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
Znt5 | |
Polyclonal | |
Unconjugated | |
SLC30A5 | |
hZTL1, SLC30A5, Zinc transporter 5, ZnT 5, ZnT like transporter 1, ZNT5, ZNTL1, ZTL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64924, 69048 | |
-20°C | |
Liquid |
Immunoprecipitation | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8R4H9, Q8TAD4 | |
SLC30A5 | |
Znt5 Fusion Protein Ag12947 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |