missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Znt5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | Znt5 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16804175
|
Proteintech
25604-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16894165
|
Proteintech
25604-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Znt5 | |
Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8R4H9, Q8TAD4 | |
64924, 69048 | |
Znt5 Fusion Protein Ag12947 | |
Primary | |
-20°C | |
SLC30A5 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
hZTL1, SLC30A5, Zinc transporter 5, ZnT 5, ZnT like transporter 1, ZNT5, ZNTL1, ZTL1 | |
SLC30A5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |