missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF593 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | ZNF593 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16898984
|
Proteintech
19426-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16888984
|
Proteintech
19426-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Negatively modulates the DNA binding activity of Oct-2 and therefore its transcriptional regulatory activity. Could act either by binding to DNA octamer or by interacting with Oct-2. May also be a modulator of other octamer-binding proteins.Specifications
| ZNF593 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| O00488, Q9DB42 | |
| 51042, 68040 | |
| ZNF593 Fusion Protein Ag13758 | |
| Primary | |
| -20°C | |
| ZNF593 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| zinc finger protein 593, Zinc finger protein T86, ZNF593, ZT86 | |
| ZNF593 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title