Learn More
ZNF593 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Product Code. 16888984 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16888984 150 μL 150µL
16898984 20 μL 20µL
2 options
Product Code. 16888984
Brand: Proteintech 194261AP150UL
Rabbit Polyclonal Antibody
Negatively modulates the DNA binding activity of Oct-2 and therefore its transcriptional regulatory activity. Could act either by binding to DNA octamer or by interacting with Oct-2. May also be a modulator of other octamer-binding proteins.
Specifications
Antigen ZNF593
Applications Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZNF593
Gene Accession No. O00488, Q9DB42
Gene Alias zinc finger protein 593, Zinc finger protein T86, ZNF593, ZT86
Gene Symbols ZNF593
Host Species Rabbit
Immunogen ZNF593 Fusion Protein Ag13758
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51042, 68040
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG