missing translation for 'onlineSavingsMsg'
Learn More

ZNF385D Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20806-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16880385

  • 4593.76 NOK / 150µL
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ZNF385D gene ontology annotations related to this gene include nucleic acid binding; zinc ion binding.
Specifications

Specifications

ZNF385D
Polyclonal
Unconjugated
ZNF385D
zinc finger protein 385D, Zinc finger protein 659, ZNF385D, ZNF659
Rabbit
Antigen Affinity Chromatography
RUO
79750
-20°C
Liquid
Western Blot
0.34 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H6B1
ZNF385D
ZNF385D Fusion Protein Ag14746
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers