missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF385D Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | ZNF385D |
---|---|
Concentration | 0.34 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890385
|
Proteintech
20806-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16880385
|
Proteintech
20806-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ZNF385D gene ontology annotations related to this gene include nucleic acid binding; zinc ion binding.Specifications
ZNF385D | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q9H6B1 | |
79750 | |
ZNF385D Fusion Protein Ag14746 | |
Primary | |
-20°C | |
ZNF385D |
0.34 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
zinc finger protein 385D, Zinc finger protein 659, ZNF385D, ZNF659 | |
ZNF385D | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |